CAS RN 85999-40-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
23-Hydroxybetulinic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
23-Hydroxybetulinic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
23-Hydroxybetulinic acid, MedChemExpress
MedChemExpress 23-hydroxybetulinic acid is one of the bioactive components responsible for its anticancer activity.