accessibility menu, dialog, popup

UserName

CH3NONHCH3

CAS RN 857283-57-9

IUPAC Name: methyl[(5-methyl-3-phenyl-1,2-oxazol-4-yl)methyl]amine
Synonyms: methyl[(5-methyl-3-phenyl-1,2-oxazol-4-yl)methyl]amine
Molecular Weight (g/mol): 202.26
Molecular Formula: C12H14N2O
InChi Key: IEMWBRRLWLUKQI-UHFFFAOYSA-N
SMILES: CNCC1=C(C)ON=C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results