CAS RN 854920-95-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,1'-Bis((2S,5S)-2,5-diisopropylphospholano)ferrocene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific™
CAS: 854920-95-9 Molecular Formula: C38H60BF4FeP2Rh Molecular Weight (g/mol): 824.40 MDL Number: MFCD15144872 SMILES: [Fe].[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC(C)[C@H]1CC[C@H](C(C)C)P1c1cccc1.CC(C)[C@H]1CC[C@H](C(C)C)P1c1cccc1