accessibility menu, dialog, popup

UserName

NNCl

CAS RN 852634-41-4

IUPAC Name: 1,3-bis(adamantan-1-yl)-3H-1λ⁵,3-benzodiazol-1-ylium chloride
Synonyms: 1,3-bis(adamantan-1-yl)-1λ⁵,3-benzodiazol-1-ylium chloride
Molecular Weight (g/mol): 423.04
Molecular Formula: C27H35ClN2
InChi Key: GMYUUHXEUIPSKA-UHFFFAOYSA-M
SMILES: [Cl-].C1C2CC3CC1CC(C2)(C3)N1C=[N+](C2=CC=CC=C12)C12CC3CC(CC(C3)C1)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results