CAS RN 850567-92-9
IUPAC Name:
(4-methylcyclohex-1-en-1-yl)boronic acid
Synonyms:
4-methylcyclohex-1-en-1-ylboronic acid
Molecular Weight (g/mol):
139.99
Molecular Formula:
C7H13BO2
InChi Key:
SJZMIZIVYIOMIW-UHFFFAOYNA-N
SMILES:
CC1CCC(=CC1)B(O)O