CAS RN 849136-14-7
CAS RN 849136-14-7
IUPAC Name:
3-bromo-N-[(pyridin-3-yl)methyl]benzene-1-sulfonamide
Synonyms:
3-bromo-N-(pyridin-3-ylmethyl)benzenesulfonamide
Molecular Weight (g/mol):
327.2
Molecular Formula:
C12H11BrN2O2S
InChi Key:
VAQOTDUYHINTAB-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC(=C1)S(=O)(=O)NCC1=CN=CC=C1
1
–
1
of
1
results
1
3-Bromo-N-(3-pyridylmethyl)benzenesulfonamide, 97%, Thermo Scientific™
CAS: 849136-14-7 Molecular Formula: C12H11BrN2O2S Molecular Weight (g/mol): 327.196 MDL Number: MFCD07356489 InChI Key: VAQOTDUYHINTAB-UHFFFAOYSA-N Synonym: 3-bromo-n-pyridin-3-ylmethyl benzenesulfonamide,3-bromo-n-3-pyridylmethyl benzenesulfonamide,3-bromo-n-pyridin-3-yl methyl benzene-1-sulfonamide PubChem CID: 3772600 IUPAC Name: 3-bromo-N-(pyridin-3-ylmethyl)benzenesulfonamide SMILES: C1=CC(=CC(=C1)Br)S(=O)(=O)NCC2=CN=CC=C2