CAS RN 847664-64-6
CAS RN 847664-64-6
IUPAC Name:
(2,5-dichloropyridin-4-yl)boronic acid
Synonyms:
2,5-dichloropyridin-4-ylboronic acid
Molecular Weight (g/mol):
191.8
Molecular Formula:
C5H4BCl2NO2
InChi Key:
BEJJMDOFMZCRST-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(Cl)=NC=C1Cl
1
–
1
of
1
results
1
2,5-Dichloropyridine-4-boronic acid, 95%, Thermo Scientific™
CAS: 847664-64-6 Molecular Formula: C5H4BCl2NO2 Molecular Weight (g/mol): 191.802 MDL Number: MFCD06798254 InChI Key: BEJJMDOFMZCRST-UHFFFAOYSA-N Synonym: 2,5-dichloropyridine-4-boronic acid,2,5-dichloropyridin-4-yl-4-boronic acid,2,5-dichloro-4-pyridyl boronic acid,2,5-dichloropyridine-4-boronicacid,2,5-dichloropyridin-4-yl boronic acid,2,5-dichloro-4-pyridineboronic acid,boronic acid, b-2,5-dichloro-4-pyridinyl,boronic acid, 2,5-dichloro-4-pyridinyl,pubchem17071,acmc-209pwe PubChem CID: 12177416 IUPAC Name: (2,5-dichloropyridin-4-yl)boronic acid SMILES: B(C1=CC(=NC=C1Cl)Cl)(O)O