CAS RN 84743-29-3
CAS RN 84743-29-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
cis-10-Pentadecenoic Acid, TRC
CAS: 84743-29-3 Molecular Formula: C15H28O2 Molecular Weight (g/mol): 240.38 Synonym: (Z)-10-Pentadecenoic Acid,(10Z)-10-Pentadecenoic Acid IUPAC Name: (Z)-pentadec-10-enoic acid SMILES: CCCC\C=C/CCCCCCCCC(=O)O