CAS RN 838-95-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(Dimethylamino)stilbene, TRC
CAS: 838-95-9 Molecular Formula: C16H17N Molecular Weight (g/mol): 223.31 SMILES: CN(C)c1ccc(\C=C\c2ccccc2)cc1