accessibility menu, dialog, popup

UserName

CAS RN 837-18-3

OSOHN

CAS RN 837-18-3

IUPAC Name: N-benzylbenzenesulfonamide
Synonyms: N-benzylbenzenesulfonamide
Molecular Weight (g/mol): 247.31
Molecular Formula: C13H13NO2S
InChi Key: GRTPAOVVVLZLDP-UHFFFAOYSA-N
SMILES: O=S(=O)(NCC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results