CAS RN 83657-24-3
CAS RN 83657-24-3
IUPAC Name:
(1E)-1-(2,4-dichlorophenyl)-4,4-dimethyl-2-(1H-1,2,4-triazol-1-yl)pent-1-en-3-ol
Synonyms:
diniconazole
Molecular Weight (g/mol):
326.22
Molecular Formula:
C15H17Cl2N3O
InChi Key:
FBOUIAKEJMZPQG-AWNIVKPZNA-N
SMILES:
CC(C)(C)C(O)C(=C/C1=CC=C(Cl)C=C1Cl)\N1C=NC=N1
1
–
5
of
5
results
2
Diniconazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Diniconazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Diniconazole, MedChemExpress
MedChemExpress Diniconazole is a newly developed fungicide strongly inhibited lanosterol 14 alpha-demethylation catalyzed by a yeast cytochrome P-450.