CAS RN 83081-75-8
IUPAC Name:
1-(2-ethoxyphenyl)piperazine hydrochloridyl
Synonyms:
1-(2-ethoxyphenyl)piperazine hydrochloridyl
Molecular Weight (g/mol):
241.74
Molecular Formula:
C12H18ClN2O
InChi Key:
RVXFSAUIRGCSJG-UHFFFAOYSA-N
SMILES:
[Cl].CCOC1=CC=CC=C1N1CCNCC1
1
–
1
of
1
results
1
1-(2-Ethoxyphenyl)piperazine Monohydrochloride 99, TRC
CAS: 83081-75-8 Molecular Formula: C12H18N2O . HCl Molecular Weight (g/mol): 242.74 Synonym: 1-(2-Ethoxyphenyl)piperazine Hydrochloride,1-(2-Ethoxyphenyl)piperazine Monohydrochloride 99 SMILES: Cl.CCOc1ccccc1N2CCNCC2