accessibility menu, dialog, popup

UserName

CAS RN 82295-98-5

CH3OOOOOCH3HO

CAS RN 82295-98-5

IUPAC Name: 4-(acetyloxy)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3-yl acetate
Synonyms: 4-(acetyloxy)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3-yl acetate
Molecular Weight (g/mol): 230.22
Molecular Formula: C10H14O6
InChi Key: LBVCUTFITQDWSU-UHFFFAOYNA-N
SMILES: CC(=O)OC1C=COC(CO)C1OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results