CAS RN 80995-97-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(-)-Isoperitenone, TRC
CAS: 80995-97-7 Molecular Formula: C10H14O Molecular Weight (g/mol): 150.22 Synonym: (-)-(4R)-Isopiperitenone,(6R)-3-Methyl-6-(1-methylethenyl)-2-cyclohexen-1-one IUPAC Name: (6R)-3-methyl-6-prop-1-en-2-ylcyclohex-2-en-1-one SMILES: CC(=C)[C@H]1CCC(=CC1=O)C