CAS RN 799264-47-4
CAS RN 799264-47-4
IUPAC Name:
1-(4-methylbenzenesulfonyl)-1H-1,3-benzodiazol-2-amine
Synonyms:
1-(4-methylbenzenesulfonyl)-1,3-benzodiazol-2-amine
Molecular Weight (g/mol):
287.34
Molecular Formula:
C14H13N3O2S
InChi Key:
SRFABRWQVPCPRG-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)S(=O)(=O)N1C(N)=NC2=CC=CC=C12
1
–
1
of
1
results
1
Nodinitib-1, MedChemExpress
MedChemExpress Nodinitib-1 (ML130;CID-1088438) is a NOD1 inhibitor with an IC50 of 0.56 μM.