CAS RN 79790-37-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dipyrazino[2,3-f:2',3'-h]quinoxaline, TRC
CAS: 79790-37-7 Molecular Formula: C12 H6 N6 Molecular Weight (g/mol): 234.216 Synonym: 1,4,5,8,9,12-Hexaazatriphenylene,Benzo[1,2-b:3,4-b':5,6-b'']tripyrazine,HAT IUPAC Name: 3,6,9,12,15,18-hexazatetracyclo[12.4.0.02,7.08,13]octadeca-1,3,5,7,9,11,13,15,17-nonaene SMILES: c1cnc2c3nccnc3c4nccnc4c2n1