CAS RN 79756-69-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
TPI-1, MedChemExpress
MedChemExpress TPI-1, also known as Tyrosine Phosphatase Inhibitor 1, is a SHP-1 inhibitor; inhibits recombinant SHP-1 with an IC50 of 40 nM.