accessibility menu, dialog, popup

UserName

CAS RN 79456-30-7

NBrH2NFFF

CAS RN 79456-30-7

IUPAC Name: 3-bromo-5-(trifluoromethyl)pyridin-2-amine
Synonyms: 3-bromo-5-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol): 241.01
Molecular Formula: C6H4BrF3N2
InChi Key: KBNACDZKKVMULE-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(C=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results