accessibility menu, dialog, popup

UserName

CH3(S)OHNHOOH3CH3CCH3

CAS RN 79069-13-9

IUPAC Name: tert-butyl N-[(2S)-1-hydroxypropan-2-yl]carbamate
Synonyms: tert-butyl N-[(2S)-1-hydroxypropan-2-yl]carbamate
Molecular Weight (g/mol): 175.23
Molecular Formula: C8H17NO3
InChi Key: PDAFIZPRSXHMCO-LURJTMIESA-N
SMILES: C[C@@H](CO)NC(=O)OC(C)(C)C
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)