accessibility menu, dialog, popup

UserName

CH3BrCH3

CAS RN 78831-87-5

IUPAC Name: 1-(bromomethyl)-2,4-dimethylbenzene
Synonyms: 1-(bromomethyl)-2,4-dimethylbenzene
Molecular Weight (g/mol): 199.09
Molecular Formula: C9H11Br
InChi Key: WGLUZJWOTTXZIC-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results