accessibility menu, dialog, popup

UserName

CAS RN 78583-82-1

NHNNH2Br

CAS RN 78583-82-1

IUPAC Name: 5-(4-bromophenyl)-1H-pyrazol-3-amine
Synonyms: 5-(4-bromophenyl)-1H-pyrazol-3-amine
Molecular Weight (g/mol): 238.09
Molecular Formula: C9H8BrN3
InChi Key: FTCWIBNIYIEHDT-UHFFFAOYSA-N
SMILES: NC1=NNC(=C1)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results