accessibility menu, dialog, popup

UserName

FHSbH3HFFHHFHF

CAS RN 7783-70-2

IUPAC Name: antimony pentafluoride
Synonyms: antimony pentafluoride
Molecular Weight (g/mol): 224.82
Molecular Formula: F5H8Sb
InChi Key: VBKPQOIVRXDIEL-UHFFFAOYSA-N
SMILES: F.F.F.F.F.[SbH3]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results