CAS RN 7779-90-0
IUPAC Name:
trizinc(2+) diphosphate
Synonyms:
zinc phosphate
Molecular Weight (g/mol):
386.08
Molecular Formula:
O8P2Zn3
InChi Key:
LRXTYHSAJDENHV-UHFFFAOYSA-H
SMILES:
[Zn++].[Zn++].[Zn++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
1
–
1
of
1
results
1
Zinc phosphate (ortho), Puratronic™, 99.995% (metals basis)
CAS: 7779-90-0 Molecular Formula: O8P2Zn3 Molecular Weight (g/mol): 386.08 MDL Number: MFCD00036282 InChI Key: LRXTYHSAJDENHV-UHFFFAOYSA-H Synonym: zinc phosphate,delaphos,trizinc diphosphate,zinc orthophosphate,trizinc phosphate,delaphos 2m,bonderite 40,granodine 80,microphos 90,granodine 16nc PubChem CID: 24519 IUPAC Name: trizinc;diphosphate SMILES: [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Zn+2].[Zn+2].[Zn+2]