CAS RN 7761-45-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Metoprine, TRC
CAS: 7761-45-7 Molecular Formula: C11 H10 Cl2 N4 Molecular Weight (g/mol): 269.13 Synonym: 5-(3,4-Dichlorophenyl)-6-methyl-2,4-pyrimidinediamine,2,4-Diamino-5-(3,4- IUPAC Name: 5-(3,4-dichlorophenyl)-6-methylpyrimidine-2,4-diamine SMILES: Cc1nc(N)nc(N)c1c2ccc(Cl)c(Cl)c2