accessibility menu, dialog, popup

UserName

CAS RN 775-16-6

ONH

CAS RN 775-16-6

IUPAC Name: 1-benzyl-3-oxopyrrolidin-1-ium
Synonyms: 1-benzyl-3-oxopyrrolidin-1-ium
Molecular Weight (g/mol): 176.24
Molecular Formula: C11H14NO
InChi Key: DHGMDHQNUNRMIN-UHFFFAOYNA-O
SMILES: O=C1CC[NH+](CC2=CC=CC=C2)C1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)