accessibility menu, dialog, popup

UserName

CAS RN 77326-36-4

NH2FN

CAS RN 77326-36-4

IUPAC Name: 2-amino-6-fluorobenzonitrile
Synonyms: 2-amino-6-fluorobenzonitrile
Molecular Weight (g/mol): 136.13
Molecular Formula: C7H5FN2
InChi Key: IQUNZGOZUJITBJ-UHFFFAOYSA-N
SMILES: NC1=CC=CC(F)=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results