CAS RN 77292-91-2
CAS RN 77292-91-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
tert-Butyl 2-(4-(Bromomethyl)phenyl)acetate, TRC
CAS: 77292-91-2 Molecular Formula: C13H17BrO2 Molecular Weight (g/mol): 285.18 IUPAC Name: tert-butyl 2-[4-(bromomethyl)phenyl]acetate SMILES: CC(C)(C)OC(=O)Cc1ccc(CBr)cc1