accessibility menu, dialog, popup

UserName

FClClFFF

CAS RN 77227-81-7

IUPAC Name: 1,3-dichloro-2-fluoro-5-(trifluoromethyl)benzene
Synonyms: 1,3-dichloro-2-fluoro-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 232.99
Molecular Formula: C7H2Cl2F4
InChi Key: BWQFQKZDLBJZAW-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=C(C=C1Cl)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results