CAS RN 7685-44-1
IUPAC Name:
(2R)-2-azaniumylpent-4-enoate
Synonyms:
allylglycine
Molecular Weight (g/mol):
115.13
Molecular Formula:
C5H9NO2
InChi Key:
WNNNWFKQCKFSDK-SCSAIBSYSA-N
SMILES:
[NH3+][C@H](CC=C)C([O-])=O
1
–
1
of
1
results
1
DL-2-Amino-4-pentenoic acid, 99%
CAS: 7685-44-1 Molecular Formula: C5H9NO2 Molecular Weight (g/mol): 115.13 MDL Number: MFCD00063103 InChI Key: WNNNWFKQCKFSDK-UHFFFAOYSA-N Synonym: dl-allylglycine,2-amino-4-pentenoic acid,dl-2-allylglycine,dl-2-amino-4-pentenoic acid,dl-c-allylglycine,4-pentenoic acid, 2-amino,dl-2-amino-1-pentenoic acid,dl-2-aminopent-4-enoic acid,s-allyl-glycine,alpha-allyl-gly-oh PubChem CID: 14044 IUPAC Name: 2-aminopent-4-enoic acid SMILES: C=CCC(C(=O)O)N