accessibility menu, dialog, popup

UserName

CAS RN 766-16-5

CH3NF

CAS RN 766-16-5

IUPAC Name: 4-fluoro-2-methylpyridine
Synonyms: 4-fluoro-2-methylpyridine
Molecular Weight (g/mol): 111.12
Molecular Formula: C6H6FN
InChi Key: LEJYGRVERQVBLB-UHFFFAOYSA-N
SMILES: CC1=NC=CC(F)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results