CAS RN 75187-63-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7,8-Dihydroxy-3-(4-hydroxy-phenyl)-chromen-4-one, TRC
CAS: 75187-63-2 Molecular Formula: C15H10O5 Molecular Weight (g/mol): 270.24 Synonym: Demethylretusin SMILES: Oc1ccc(cc1)C2=COc3c(O)c(O)ccc3C2=O