CAS RN 75181-07-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
332.8
Molecular Formula:
C10H15Cl3Zr
InChi Key:
XZRIJZMMTDVZIR-UHFFFAOYSA-K
SMILES:
Cl[Zr](Cl)Cl.Cc1c(C)c(C)[c-](C)c1C
1
–
1
of
1
results
1
Pentamethylcyclopentadienylzirconium trichloride, 99%, Thermo Scientific Chemicals
CAS: 75181-07-6 MDL Number: MFCD00070444