accessibility menu, dialog, popup

UserName

CAS RN 7502-06-9

H3CONH2

CAS RN 7502-06-9

IUPAC Name: 4-(hexadecyloxy)aniline
Synonyms: 4-(hexadecyloxy)aniline
Molecular Weight (g/mol): 333.56
Molecular Formula: C22H39NO
InChi Key: DETPMDIIFZGDAE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCOC1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results