accessibility menu, dialog, popup

UserName

CAS RN 7465-94-3

H3COONHClCl

CAS RN 7465-94-3

IUPAC Name: N-(2,5-dichlorophenyl)-4-methoxybenzamide
Synonyms: N-(2,5-dichlorophenyl)-4-methoxybenzamide
Molecular Weight (g/mol): 296.15
Molecular Formula: C14H11Cl2NO2
InChi Key: XQSFQIDFRWNBTM-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)NC1=CC(Cl)=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results