accessibility menu, dialog, popup

UserName

CAS RN 745784-03-6

IUPAC Name: 3,5-difluoropyridine-2-carboxamide
Synonyms: 3,5-difluoropyridine-2-carboxamide
Molecular Weight (g/mol): 158.11
Molecular Formula: C6H4F2N2O
InChi Key: LWKSZYXOBLMONP-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(F)C=C(F)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results