CAS RN 74274-36-5
CAS RN 74274-36-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Acifluorfen-2-amino, TRC
CAS: 74274-36-5 Molecular Formula: C14 H9 Cl F3 N O3 Molecular Weight (g/mol): 331.67 Synonym: 2-Amino-5-[2-chloro-4-(trifluoromethyl)phenoxy]benzoic acid,4-Amino-3-carboxy-2'-chloro-4'-trifluoromethyldiphenyl ether IUPAC Name: 2-amino-5-[2-chloro-4-(trifluoromethyl)phenoxy]benzoic acid SMILES: Nc1ccc(Oc2ccc(cc2Cl)C(F)(F)F)cc1C(=O)O