accessibility menu, dialog, popup

UserName

CAS RN 73953-89-6

IUPAC Name: 1-benzyl-1H-1,2,3-triazole-4,5-dicarboxylate
Synonyms: 1-benzyl-1,2,3-triazole-4,5-dicarboxylate
Molecular Weight (g/mol): 245.2
Molecular Formula: C11H7N3O4
InChi Key: IYYBUXUZYWGZGB-UHFFFAOYSA-L
SMILES: [O-]C(=O)C1=C(N(CC2=CC=CC=C2)N=N1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results