accessibility menu, dialog, popup

UserName

CH3H3CCH3OON

CAS RN 73286-70-1

IUPAC Name: tert-butyl 2,5-dihydro-1H-pyrrole-1-carboxylate
Synonyms: tert-butyl 2,5-dihydropyrrole-1-carboxylate
Molecular Weight (g/mol): 169.22
Molecular Formula: C9H15NO2
InChi Key: YEBDZDMYLQHGGZ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CC=CC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)