CAS RN 729582-16-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-[(1,1-Dioxo-1,2-benzothiazol-3-yl)amino]hexanoic Acid, TRC
CAS: 729582-16-5 Molecular Formula: C13H16N2O4S Molecular Weight (g/mol): 296.34 SMILES: OC(=O)CCCCCNC1=NS(=O)(=O)c2ccccc12