CAS RN 728-81-4
IUPAC Name:
phenyl[3-(trifluoromethyl)phenyl]methanone
Synonyms:
phenyl[3-(trifluoromethyl)phenyl]methanone
Molecular Weight (g/mol):
250.22
Molecular Formula:
C14H9F3O
InChi Key:
IOXDAYKKVHAKSX-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
3-(Trifluoromethyl)benzophenone, 97+%, Thermo Scientific™
CAS: 728-81-4 Molecular Formula: C14H9F3O Molecular Weight (g/mol): 250.22 MDL Number: MFCD00000389 InChI Key: IOXDAYKKVHAKSX-UHFFFAOYSA-N Synonym: 3-trifluoromethyl benzophenone,phenyl 3-trifluoromethyl phenyl methanone,m-trifluoromethylbenzophenone,phenyl-3-trifluoromethyl-phenyl-methanone,phenyl-3-trifluoromethyl phenyl methanone,fxffr cvr,phenyl 3-trifluoromethyl phenyl ketone,pubchem1923,zlchem 1027 PubChem CID: 69766 IUPAC Name: phenyl-[3-(trifluoromethyl)phenyl]methanone SMILES: FC(F)(F)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1