CAS RN 72762-00-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Levetiracetam EP Impurity C, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2-Hydroxypyridine, TRC
CAS: 72762-00-6 Molecular Formula: C5 H5 N O Molecular Weight (g/mol): 95.1 Synonym: Pyridin-2-ol,Levetiracetam Imp. C (EP),2(1H)-Pyridone (6CI,8CI),2-Pyridol (3CI),1,2-Dihydro-2-oxopyridine,1H-2-Pyridone,1H-Pyridin-2-one,2-Hydroxypyridine,2-Oxopyridine,2-Pyridinol,2-Pyridinone,2-Pyridone,NSC 172522,Pyridin-2-one,α-Hydroxypyridine,α-Pyridone IUPAC Name: pyridin-2-ol SMILES: Oc1ccccn1