CAS RN 725-05-3
CAS RN 725-05-3
IUPAC Name:
4'-methoxy-[1,1'-biphenyl]-3-carboxylate
Synonyms:
4'-methoxy-[1,1'-biphenyl]-3-carboxylate
Molecular Weight (g/mol):
227.24
Molecular Formula:
C14H11O3
InChi Key:
VWDMBLJBLIUXFS-UHFFFAOYSA-M
SMILES:
COC1=CC=C(C=C1)C1=CC=CC(=C1)C([O-])=O
1
–
1
of
1
results
1
4'-Methoxybiphenyl-3-carboxylic acid, 95%, Thermo Scientific™
CAS: 725-05-3 Molecular Formula: C14H12O3 Molecular Weight (g/mol): 228.247 MDL Number: MFCD03424599 InChI Key: VWDMBLJBLIUXFS-UHFFFAOYSA-N Synonym: 4'-methoxy-1,1'-biphenyl-3-carboxylic acid,3-4-methoxyphenyl benzoic acid,4'-methoxybiphenyl-3-carboxylic acid,4'-methoxy-biphenyl-3-carboxylic acid,4'-methoxy 1,1'-biphenyl-3-carboxylic acid,3-biphenyl-4'-methoxy carboxylic acid,1,1'-biphenyl-3-carboxylic acid, 4'-methoxy,acmc-1bff1,amtda012,4'-methoxy-3-biphenylcarboxylic acid PubChem CID: 1307667 IUPAC Name: 3-(4-methoxyphenyl)benzoic acid SMILES: COC1=CC=C(C=C1)C2=CC(=CC=C2)C(=O)O