CAS RN 72301-79-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Enviroxime, TRC
CAS: 72301-79-2 Molecular Formula: C17 H18 N4 O3 S Molecular Weight (g/mol): 358.41 Synonym: 6-[(E)-(Hydroxyimino)phenylmethyl]-1-[(1-methylethyl)sulfonyl]-1H-benzimidazol-2-amine,LY 122772 IUPAC Name: (NE)-N-[(2-amino-3-propan-2-ylsulfonylbenzimidazol-5-yl)-phenylmethylidene]hydroxylamine SMILES: CC(C)S(=O)(N1C2=CC(/C(C3=CC=CC=C3)=N/O)=CC=C2N=C1N)=O