CAS RN 72136-45-9
CAS RN 72136-45-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
[(N-Acryloylamino)phenyl]mercuric Chloride, 90%, TRC
CAS: 72136-45-9 Molecular Formula: C9H8ClHgNO Molecular Weight (g/mol): 382.21 Synonym: Chloro[4-[(1-oxo-2-propen-1-yl)amino]phenyl]mercury,N-Phenyl-2-propenamide Mercury Complex,APM; IUPAC Name: chloro-[4-(prop-2-enoylamino)phenyl]mercury SMILES: C=CC(NC1=CC=C([Hg]Cl)C=C1)=O