CAS RN 7146-68-1
CAS RN 7146-68-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Amino-4'-chlorodiphenyl Sulfone, TRC
CAS: 7146-68-1 Molecular Formula: C12 H10 Cl N O2 S Molecular Weight (g/mol): 267.73 Synonym: 4-[(4-Chlorophenyl)sulfonyl]benzenamine,p-[(p-Chlorophenyl)sulfonyl]aniline,4-(4-Chlorobenzenesulfonyl)phenylamine,4-Amino-4'-chlorodiphenyl Sulfone,4-Aminophenyl 4-chlorophenyl Sulfone,NSC 23812,NSC 51990,p-(p-Chlorophenylsulfonyl)aniline,p-Aminophenyl p-Chlorophenyl Sulfone; IUPAC Name: 4-(4-chlorophenyl)sulfonylaniline SMILES: Nc1ccc(cc1)S(=O)(=O)c2ccc(Cl)cc2