CAS RN 71414-47-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Pentamethylcyclopentadienyltantalum tetrachloride, Thermo Scientific™
CAS: 71414-47-6 MDL Number: MFCD00070463