accessibility menu, dialog, popup

UserName

CH3OOH2NHCl

CAS RN 712313-64-9

IUPAC Name: methyl 3-aminocyclohexane-1-carboxylate hydrochloride
Synonyms: methyl 3-aminocyclohexane-1-carboxylate hydrochloride
Molecular Weight (g/mol): 193.67
Molecular Formula: C8H16ClNO2
InChi Key: OOFXENVWECZJBF-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C1CCCC(N)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results