CAS RN 70806-55-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
trans-Tranilast, MedChemExpress
MedChemExpress trans-Tranilast (trans-MK-341) is an antiallergic drug, used to treat bronchial asthma, allergic rhinitis and atopic dermatitis.