accessibility menu, dialog, popup

UserName

CH3NOO

CAS RN 70753-36-5

IUPAC Name: 3-methyl-1,2-oxazole-5-carbaldehyde
Synonyms: 3-methyl-1,2-oxazole-5-carbaldehyde
Molecular Weight (g/mol): 111.1
Molecular Formula: C5H5NO2
InChi Key: GEWPFPUABRRBPS-UHFFFAOYSA-N
SMILES: CC1=NOC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results