CAS RN 70201-42-2
IUPAC Name:
3,5-dibromopyridine-4-carbaldehyde
Synonyms:
3,5-dibromopyridine-4-carbaldehyde
Molecular Weight (g/mol):
264.9
Molecular Formula:
C6H3Br2NO
InChi Key:
WPBYVMDYYFWYAY-UHFFFAOYSA-N
SMILES:
BrC1=CN=CC(Br)=C1C=O
1
–
1
of
1
results
1
3,5-Dibromoisonicotinaldehyde, TRC
CAS: 70201-42-2 Molecular Formula: C6H3NOBr2 Molecular Weight (g/mol): 264.9 IUPAC Name: 3,5-dibromopyridine-4-carbaldehyde SMILES: Brc1cncc(Br)c1C=O